Product Details

SNP ID
rs150922490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140525544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGCGAATATTTGATACCTATG[G/T]AGGTGCACAAAGGACACACAGGATA
Phenotype
MIM: 610196
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ELMOD2 PubMed Links

Gene Details

Gene
ELMOD2
Gene Name
ELMO domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153702.3 605 Missense Mutation GGA,GTA G39V NP_714913.1
XM_005262885.3 605 Missense Mutation GGA,GTA G51V XP_005262942.1
XM_005262886.2 605 Missense Mutation GGA,GTA G39V XP_005262943.1
XM_011531818.2 605 Missense Mutation GGA,GTA G51V XP_011530120.1
XM_011531819.2 605 Missense Mutation GGA,GTA G39V XP_011530121.1

View Full Product Details