Product Details

SNP ID
rs150960276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:42401040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTACGTCCCCTTTCTCATCGT[C/T]GGCTCCATCTTCATTGCGTTCATCA
Phenotype
MIM: 609542
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8A1 PubMed Links

Gene Details

Gene
ATP8A1
Gene Name
ATPase phospholipid transporting 8A1
There are no transcripts associated with this gene.

Gene
LOC105374428
Gene Name
uncharacterized LOC105374428
There are no transcripts associated with this gene.

Gene
SHISA3
Gene Name
shisa family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080505.1 524 Silent Mutation GTC,GTT V102V NP_001073974.1

View Full Product Details