Product Details

SNP ID
rs150973681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186588871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGTTTGAGGTTCAGACATATC[A/G]AGGGGATAAAAATTGGGCAAATACT
Phenotype
MIM: 600976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAT1 PubMed Links

Gene Details

Gene
FAT1
Gene Name
FAT atypical cadherin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005245.3 13676 Silent Mutation CTC,CTT L4496L NP_005236.2
XM_005262834.2 13676 Silent Mutation CTC,CTT L4508L XP_005262891.1
XM_005262835.2 13676 Silent Mutation CTC,CTT L4508L XP_005262892.1
XM_006714139.2 13676 Silent Mutation CTC,CTT L4496L XP_006714202.1

View Full Product Details