Product Details

SNP ID
rs151038686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82819146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGAAAGCAGAGGTCTCACTGAA[A/G]TTGCTGGTGCTAACTATGTGGGTAT
Phenotype
MIM: 610257
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC31A PubMed Links

Gene Details

Gene
SEC31A
Gene Name
SEC31 homolog A, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077206.3 3671 Silent Mutation AAC,AAT N1083N NP_001070674.1
NM_001077207.3 3671 Silent Mutation AAC,AAT N1197N NP_001070675.1
NM_001077208.3 3671 Silent Mutation AAC,AAT N1182N NP_001070676.1
NM_001191049.2 3671 Silent Mutation AAC,AAT N1177N NP_001177978.1
NM_001300744.2 3671 Silent Mutation AAC,AAT N1044N NP_001287673.1
NM_001300745.2 3671 Silent Mutation AAC,AAT N1143N NP_001287674.1
NM_001318119.1 3671 Silent Mutation AAC,AAT N1182N NP_001305048.1
NM_001318120.1 3671 Silent Mutation AAC,AAT N1197N NP_001305049.1
NM_016211.4 3671 Silent Mutation AAC,AAT N1158N NP_057295.2

View Full Product Details