Product Details

SNP ID
rs151044158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:52051728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGGCGGAAAACCTGAAAAGACT[C/G]GTCTCAAACGAAACTTTACGAACGT
Phenotype
MIM: 612814
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPATA18 PubMed Links

Gene Details

Gene
SPATA18
Gene Name
spermatogenesis associated 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297608.1 313 Silent Mutation CTC,CTG L8L NP_001284537.1
NM_145263.3 313 Silent Mutation CTC,CTG L8L NP_660306.1
XM_005265716.1 313 UTR 5 XP_005265773.1
XM_005265717.2 313 Silent Mutation CTC,CTG L8L XP_005265774.1
XM_005265718.1 313 UTR 5 XP_005265775.1
XM_011534369.1 313 UTR 5 XP_011532671.1
XM_011534370.1 313 UTR 5 XP_011532672.1
XM_011534372.1 313 UTR 5 XP_011532674.1
XM_017007731.1 313 Intron XP_016863220.1

View Full Product Details