Product Details

SNP ID
rs151238403
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76306906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGAAAGACGCCCCTCTTGGGG[A/G]AGCTGCGATTCCGGGAGGCCATCAG
Phenotype
MIM: 607406
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC158 PubMed Links

Gene Details

Gene
CCDC158
Gene Name
coiled-coil domain containing 158
There are no transcripts associated with this gene.

Gene
FAM47E-STBD1
Gene Name
FAM47E-STBD1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242939.1 881 Intron NP_001229868.1
Gene
STBD1
Gene Name
starch binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003943.4 881 Missense Mutation GAA,GGA E46G NP_003934.1

View Full Product Details