Product Details

SNP ID
rs151269794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:146707546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCTTCCACTGTGGATCCTCTAA[A/T]CTCTTCTAAAAAAAAAATACACAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TTC29 PubMed Links

Gene Details

Gene
TTC29
Gene Name
tetratricopeptide repeat domain 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300761.2 1668 Missense Mutation ATT,TTT I472F NP_001287690.1
NM_001317806.1 1668 Missense Mutation ATT,TTT I445F NP_001304735.1
NM_031956.3 1668 Missense Mutation ATT,TTT I446F NP_114162.2
XM_005263270.1 1668 Missense Mutation ATT,TTT I463F XP_005263327.1
XM_006714335.1 1668 Missense Mutation ATT,TTT I471F XP_006714398.1
XM_006714336.1 1668 Missense Mutation ATT,TTT I462F XP_006714399.1
XM_006714339.2 1668 Missense Mutation ATT,TTT I410F XP_006714402.1
XM_011532310.2 1668 Intron XP_011530612.1

View Full Product Details