Product Details

SNP ID
rs151277844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56979002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGCAATATGTACGACGCGGATG[A/G]GGGTAGGTGAACGCTCAAAACACAC
Phenotype
MIM: 614919 MIM: 180661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOA1 PubMed Links

Gene Details

Gene
NOA1
Gene Name
nitric oxide associated 1
There are no transcripts associated with this gene.

Gene
POLR2B
Gene Name
RNA polymerase II subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000938.2 363 Missense Mutation GAG,GGG E6G NP_000929.1
NM_001303268.1 363 UTR 5 NP_001290197.1
NM_001303269.1 363 UTR 5 NP_001290198.1

View Full Product Details