Product Details
- SNP ID
-
rs151298217
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:20732705 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGAAAAAATGTTTCAACGTGTTG[C/T]CTGGGAGCATCTTCTTTGAGGACAG
- Phenotype
-
MIM: 608182
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KCNIP4
PubMed Links
Gene Details
- Gene
- KCNIP4
- Gene Name
- potassium voltage-gated channel interacting protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001035003.1 |
651 |
Silent Mutation |
AGA,AGG |
R181R |
NP_001030175.1 |
NM_001035004.1 |
651 |
Silent Mutation |
AGA,AGG |
R144R |
NP_001030176.1 |
NM_025221.5 |
651 |
Silent Mutation |
AGA,AGG |
R206R |
NP_079497.2 |
NM_147181.3 |
651 |
Silent Mutation |
AGA,AGG |
R172R |
NP_671710.1 |
NM_147182.3 |
651 |
Silent Mutation |
AGA,AGG |
R144R |
NP_671711.1 |
NM_147183.3 |
651 |
Silent Mutation |
AGA,AGG |
R185R |
NP_671712.1 |
XM_005248190.4 |
651 |
Silent Mutation |
AGA,AGG |
R189R |
XP_005248247.1 |
XM_011513882.2 |
651 |
Silent Mutation |
AGA,AGG |
R206R |
XP_011512184.1 |
XM_011513883.2 |
651 |
Silent Mutation |
AGA,AGG |
R189R |
XP_011512185.1 |
XM_011513884.2 |
651 |
Silent Mutation |
AGA,AGG |
R185R |
XP_011512186.1 |
XM_011513885.2 |
651 |
Silent Mutation |
AGA,AGG |
R181R |
XP_011512187.1 |
XM_011513886.2 |
651 |
Silent Mutation |
AGA,AGG |
R172R |
XP_011512188.1 |
XM_011513887.2 |
651 |
Silent Mutation |
AGA,AGG |
R144R |
XP_011512189.1 |
XM_011513889.2 |
651 |
Silent Mutation |
AGA,AGG |
R144R |
XP_011512191.1 |
XM_017008653.1 |
651 |
Silent Mutation |
AGA,AGG |
R144R |
XP_016864142.1 |
- Gene
- PACRGL
- Gene Name
- PARK2 coregulated like
View Full Product Details