Product Details

SNP ID
rs121912286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:51389922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAACTCCAGCAGCAGCAGCCCA[A/G]TGACAAAACTGTGAGTGGCTCTGGG
Phenotype
MIM: 600366
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISL1 PubMed Links

Gene Details

Gene
ISL1
Gene Name
ISL LIM homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002202.2 1303 Missense Mutation AAT,AGT N252S NP_002193.2
XM_011543380.2 1303 Missense Mutation AAT,AGT N188S XP_011541682.1
Gene
LOC642366
Gene Name
uncharacterized LOC642366
There are no transcripts associated with this gene.

View Full Product Details