Product Details

SNP ID
rs137901435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170105987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTTCGACCTGCCGGCGCCCTC[C/T]CCACCTCGCTGCAGCCCCCAGTTCC
Phenotype
MIM: 601093
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXI1 PubMed Links

Gene Details

Gene
FOXI1
Gene Name
forkhead box I1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012188.4 75 Silent Mutation TCC,TCT S10S NP_036320.2
NM_144769.2 75 Silent Mutation TCC,TCT S10S NP_658982.1

View Full Product Details