Product Details

SNP ID
rs137995316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132866891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCCGCCGCCACAATGGGCCGC[A/G]AGTTTGGGAATCTGACGCGGATGCG
Phenotype
MIM: 604417 MIM: 601918 MIM: 611373 MIM: 612080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFF4 PubMed Links

Gene Details

Gene
AFF4
Gene Name
AF4/FMR2 family member 4
There are no transcripts associated with this gene.

Gene
GDF9
Gene Name
growth differentiation factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288824.2 84 Intron NP_001275753.1
NM_001288825.2 84 Intron NP_001275754.1
NM_001288826.2 84 Intron NP_001275755.1
NM_001288827.2 84 Intron NP_001275756.1
NM_001288828.2 84 Intron NP_001275757.1
XM_005271957.4 84 Intron XP_005272014.1
XM_006714585.3 84 Intron XP_006714648.1
XM_011543308.2 84 Intron XP_011541610.1
XM_011543309.1 84 Intron XP_011541611.1
XM_011543310.1 84 Intron XP_011541612.1
XM_011543311.2 84 Intron XP_011541613.1
Gene
LEAP2
Gene Name
liver enriched antimicrobial peptide 2
There are no transcripts associated with this gene.

Gene
UQCRQ
Gene Name
ubiquinol-cytochrome c reductase complex III subunit VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014402.4 84 Missense Mutation AAG,GAG K4E NP_055217.2

View Full Product Details