Product Details

SNP ID
rs138122219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:144180594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGTAGCTCATCTCTGTTCCA[A/C]ACAACTTCAGCTGGGGAGCTCAAAA
Phenotype
MIM: 613423 MIM: 611483
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCTD16 PubMed Links

Gene Details

Gene
KCTD16
Gene Name
potassium channel tetramerization domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020768.3 Intron NP_065819.1
XM_005268492.2 Intron XP_005268549.1
XM_005268493.2 Intron XP_005268550.1
XM_006714789.2 Intron XP_006714852.1
XM_011537671.2 Intron XP_011535973.1
XM_011537672.2 Intron XP_011535974.1
XM_011537673.2 Intron XP_011535975.1
Gene
YIPF5
Gene Name
Yip1 domain family member 5
There are no transcripts associated with this gene.

View Full Product Details