Product Details

SNP ID
rs138390777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1201695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCCCGGCCTAGACGCCCGGAT[C/G]CCGTCCCTGGCTGAGCTGGAGACCA
Phenotype
MIM: 608893
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC6A19 PubMed Links

Gene Details

Gene
SLC6A19
Gene Name
solute carrier family 6 member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003841.2 101 Missense Mutation ATC,ATG I15M NP_001003841.1

View Full Product Details