Product Details

SNP ID
rs138433207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:96393173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGGGCTTCGTAGAAGTTTTCA[G/T]AAGGGATGTTGAGCTTTGCACTTGG
Phenotype
MIM: 162150
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929710 PubMed Links

Gene Details

Gene
LOC101929710
Gene Name
uncharacterized LOC101929710
There are no transcripts associated with this gene.

Gene
PCSK1
Gene Name
proprotein convertase subtilisin/kexin type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000439.4 2329 Missense Mutation TAT,TCT Y697S NP_000430.3
NM_001177875.1 2329 Missense Mutation TAT,TCT Y650S NP_001171346.1

View Full Product Details