Product Details

SNP ID
rs138440180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55231638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGCCAGACTCAGCTCTGCCACCC[C/G]CCGCGCCAGGGCTTGCGCTTCCAGA
Phenotype
MIM: 607752 MIM: 614086
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCNO PubMed Links

Gene Details

Gene
CCNO
Gene Name
cyclin O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021147.4 984 Missense Mutation CGG,GGG R264G NP_066970.3
Gene
MCIDAS
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
There are no transcripts associated with this gene.

View Full Product Details