Product Details

SNP ID
rs138563518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159270543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCATTAGGGAAGAAAACCTACT[G/T]AAAATTTCTCGCAGAGTGAAAGAGT
Phenotype
MIM: 609867
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBLCP1 PubMed Links

Gene Details

Gene
UBLCP1
Gene Name
ubiquitin like domain containing CTD phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145049.3 674 Silent Mutation CTG,CTT L116L NP_659486.2

View Full Product Details