Product Details

SNP ID
rs139029027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140673319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCCCGTTCACCTGCACCAGGC[C/G]GATGCCTGTCTCCCTGACCCACGCC
Phenotype
MIM: 609385 MIM: 142810
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DND1 PubMed Links

Gene Details

Gene
DND1
Gene Name
DND microRNA-mediated repression inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194249.2 3353 Intron NP_919225.1
Gene
HARS
Gene Name
histidyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258040.2 3353 Intron NP_001244969.1
NM_001258041.2 3353 Intron NP_001244970.1
NM_001258042.2 3353 Intron NP_001244971.1
NM_001289092.1 3353 Intron NP_001276021.1
NM_001289093.1 3353 Intron NP_001276022.1
NM_001289094.1 3353 Intron NP_001276023.1
NM_002109.5 3353 Intron NP_002100.2
Gene
WDR55
Gene Name
WD repeat domain 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017706.4 3353 Intron NP_060176.2
XM_005268469.2 3353 UTR 3 XP_005268526.1
XM_017009600.1 3353 UTR 3 XP_016865089.1

View Full Product Details