Product Details
- SNP ID
-
rs139061980
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:180102113 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTTTTTCTGTTTGGTTCTCTGGG[C/T]TCTCACTTTCGTAAGAAGCCAAATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RASGEF1C
PubMed Links
Gene Details
- Gene
- RASGEF1C
- Gene Name
- RasGEF domain family member 1C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175062.3 |
1443 |
Missense Mutation |
AAC,AGC |
N445S |
NP_778232.2 |
View Full Product Details