Product Details

SNP ID
rs139081093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141330846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACATCGCCAAGGACCTAGGGCT[A/G]CAACCCCAGGAGCTGGCAGATGGCG
Phenotype
MIM: 606288 MIM: 606289
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 162 Silent Mutation CTA,CTG L54L NP_061735.1
NM_031993.1 162 Silent Mutation CTA,CTG L54L NP_114382.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
There are no transcripts associated with this gene.

View Full Product Details