Product Details
- SNP ID
-
rs139105148
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:97096833 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGTCGGGAACACTTTGTTTCAC[A/G]AAGGGCTTTTAGGCTTCCATTCCAG
- Phenotype
-
MIM: 610466
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LIX1
PubMed Links
Gene Details
- Gene
- LIX1
- Gene Name
- limb and CNS expressed 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153234.4 |
778 |
Missense Mutation |
CGT,TGT |
R180C |
NP_694966.3 |
View Full Product Details