Product Details

SNP ID
rs139302531
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:14710284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTTCAGTTCACTGTAAAAACT[A/G]AAATGCAAAGTTAGGATGCTCCATG
Phenotype
MIM: 605145 MIM: 615712
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANKH PubMed Links
Additional Information
For this assay, SNP(s) [rs71603731] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ANKH
Gene Name
ANKH inorganic pyrophosphate transport regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054027.4 2723 UTR 3 NP_473368.1
XM_011514067.1 2723 Intron XP_011512369.1
XM_017009644.1 2723 UTR 3 XP_016865133.1
Gene
LOC100130744
Gene Name
uncharacterized LOC100130744
There are no transcripts associated with this gene.

Gene
OTULIN
Gene Name
OTU deubiquitinase with linear linkage specificity
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138348.4 2723 Intron NP_612357.4
XM_011514151.2 2723 Intron XP_011512453.1
XM_011514152.2 2723 Intron XP_011512454.1
XM_011514154.2 2723 Intron XP_011512456.1
XM_017010015.1 2723 Intron XP_016865504.1

View Full Product Details