Product Details

SNP ID
rs139329842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141094883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCATAGCTCAGGCTAGTTGCCA[G/T]CCTAGGCACTATTCAGTGGCCGAGG
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018936.3 277 Missense Mutation CAG,CAT Q31H NP_061759.1
Gene
PCDHB3
Gene Name
protocadherin beta 3
There are no transcripts associated with this gene.

View Full Product Details