Product Details

SNP ID
rs139518432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73123503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGTCCTGCTCTCCATCTTTGGG[C/T]TCCAGGCATGCCAATATAAGCCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105379030 PubMed Links

Gene Details

Gene
LOC105379030
Gene Name
uncharacterized LOC105379030
There are no transcripts associated with this gene.

Gene
TMEM171
Gene Name
transmembrane protein 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161342.2 334 Missense Mutation CTC,TTC L44F NP_001154814.1
NM_173490.7 334 Missense Mutation CTC,TTC L44F NP_775761.4
XM_011543156.1 334 Missense Mutation CTC,TTC L44F XP_011541458.1

View Full Product Details