Product Details

SNP ID
rs139629564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34936198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATGATTGCCAAGGAAGAACTA[C/G]AATCTGTGTTAGAGGAAGAGGTTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC21 PubMed Links

Gene Details

Gene
DNAJC21
Gene Name
DnaJ heat shock protein family (Hsp40) member C21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012339.2 597 Missense Mutation CAA,GAA Q124E NP_001012339.2
NM_194283.3 597 Missense Mutation CAA,GAA Q124E NP_919259.3
XM_005248249.4 597 Missense Mutation CAA,GAA Q211E XP_005248306.2
XM_005248250.3 597 Missense Mutation CAA,GAA Q211E XP_005248307.2
XM_011513965.2 597 Missense Mutation CAA,GAA Q211E XP_011512267.2
XM_011513966.2 597 Missense Mutation CAA,GAA Q211E XP_011512268.2

View Full Product Details