Product Details
- SNP ID
-
rs139900667
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:172209709 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACCCTACATACTAAAATTATACA[C/T]ATCAAGTACTGGTCCAATCTTATAT
- Phenotype
-
MIM: 610174
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFCAB9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3836824] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFCAB9
- Gene Name
- EF-hand calcium binding domain 9
There are no transcripts associated with this gene.
- Gene
- UBTD2
- Gene Name
- ubiquitin domain containing 2
View Full Product Details