Product Details

SNP ID
rs140038439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139477491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAACAAAGTCTGCAAGGGGGTGG[C/T]GTACTCCAGGACACAGGTGCCCGCC
Phenotype
MIM: 612374
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM173 PubMed Links

Gene Details

Gene
TMEM173
Gene Name
transmembrane protein 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301738.1 821 Intron NP_001288667.1
NM_198282.3 821 Missense Mutation ACC,GCC T262A NP_938023.1
XM_005268445.3 821 Intron XP_005268502.1
XM_011537639.2 821 Intron XP_011535941.1
XM_011537640.2 821 Missense Mutation ACC,GCC T143A XP_011535942.1

View Full Product Details