Product Details
- SNP ID
-
rs140324183
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:36253665 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTACCTGAATTAAAAATATTTT[A/G]ATGCTATAGTCTTCTAAATCAGTAG
- Phenotype
-
MIM: 616391
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RANBP3L
PubMed Links
Gene Details
- Gene
- RANBP3L
- Gene Name
- RAN binding protein 3 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001161429.2 |
1575 |
Silent Mutation |
ATC,ATT |
I408I |
NP_001154901.1 |
NM_001323273.1 |
1575 |
Silent Mutation |
ATC,ATT |
I383I |
NP_001310202.1 |
NM_001323274.1 |
1575 |
Silent Mutation |
ATC,ATT |
I408I |
NP_001310203.1 |
NM_001323275.1 |
1575 |
Silent Mutation |
ATC,ATT |
I388I |
NP_001310204.1 |
NM_001323276.1 |
1575 |
Silent Mutation |
ATC,ATT |
I388I |
NP_001310205.1 |
NM_001323277.1 |
1575 |
Silent Mutation |
ATC,ATT |
I383I |
NP_001310206.1 |
NM_001323278.1 |
1575 |
Silent Mutation |
ATC,ATT |
I363I |
NP_001310207.1 |
NM_001323279.1 |
1575 |
Silent Mutation |
ATC,ATT |
I359I |
NP_001310208.1 |
NM_001323280.1 |
1575 |
Silent Mutation |
ATC,ATT |
I363I |
NP_001310209.1 |
NM_145000.4 |
1575 |
Silent Mutation |
ATC,ATT |
I383I |
NP_659437.3 |
XM_006714452.3 |
1575 |
Silent Mutation |
ATC,ATT |
I408I |
XP_006714515.1 |
XM_006714453.3 |
1575 |
Silent Mutation |
ATC,ATT |
I388I |
XP_006714516.1 |
XM_006714455.3 |
1575 |
Silent Mutation |
ATC,ATT |
I363I |
XP_006714518.1 |
View Full Product Details