Product Details

SNP ID
rs140336081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:142311414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAGGTCAAGCCGAGGCATCGT[C/G]CAGCCAAACAGGCCATTGGTGACAG
Phenotype
MIM: 607984
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPRY4 PubMed Links

Gene Details

Gene
SPRY4
Gene Name
sprouty RTK signaling antagonist 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127496.1 4005 UTR 3 NP_001120968.1
NM_001293289.1 4005 UTR 3 NP_001280218.1
NM_001293290.1 4005 UTR 3 NP_001280219.1
NM_030964.3 4005 UTR 3 NP_112226.2
XM_011537685.2 4005 UTR 3 XP_011535987.1
XM_017009910.1 4005 UTR 3 XP_016865399.1
Gene
SPRY4-IT1
Gene Name
SPRY4 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details