Product Details
- SNP ID
-
rs140420820
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:148827076 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTGGCCTGTGCTGATCTGGTCA[A/T]GGGCCTGGCAGTGGTGCCCTTTGGG
- Phenotype
-
MIM: 109690
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADRB2
PubMed Links
Gene Details
- Gene
- ADRB2
- Gene Name
- adrenoceptor beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000024.5 |
484 |
Missense Mutation |
AAG,ATG |
K82M |
NP_000015.1 |
View Full Product Details