Product Details

SNP ID
rs140606584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172768817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGGTAGCTCAGCGCACTGTTC[A/G]TGGAGTGGACAGGGATGGAGACGGG
Phenotype
MIM: 600714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP1 PubMed Links

Gene Details

Gene
DUSP1
Gene Name
dual specificity phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004417.3 1297 Missense Mutation ACG,ATG T350M NP_004408.1
Gene
LOC101928093
Gene Name
uncharacterized LOC101928093
There are no transcripts associated with this gene.

View Full Product Details