Product Details

SNP ID
rs140696000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37396538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAATGAAGAAGAAGAAGAAGCA[A/G]AGGAAGAAGAAGAGGAAGAGGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR70 PubMed Links

Gene Details

Gene
WDR70
Gene Name
WD repeat domain 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018034.2 536 Missense Mutation AAG,GAG K154E NP_060504.1
XM_005248318.3 536 Missense Mutation AAG,GAG K153E XP_005248375.1
XM_011514061.2 536 Intron XP_011512363.1
XM_017009604.1 536 Intron XP_016865093.1
XM_017009605.1 536 Intron XP_016865094.1

View Full Product Details