Product Details

SNP ID
rs140793388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138252978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGGAAGTCCAGCTACCATAGG[C/T]TCAGGAGCAGAATCAAGGGAAGCGT
Phenotype
MIM: 605710
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFRA3 PubMed Links

Gene Details

Gene
GFRA3
Gene Name
GDNF family receptor alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001496.3 1333 Missense Mutation AAC,AGC N398S NP_001487.2

View Full Product Details