Product Details

SNP ID
rs140911228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157162890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAAGACGCTTTCTGATTCTTGT[C/T]CCCCGCTCTCCTGCGGTGACTTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM71B PubMed Links

Gene Details

Gene
FAM71B
Gene Name
family with sequence similarity 71 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130899.2 1475 Missense Mutation NP_570969.2

View Full Product Details