Product Details

SNP ID
rs141047933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:147826105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGTTTATGGAAAAGAATTTGC[C/T]GGTACAGATTCCCTTTGAAGACGTA
Phenotype
MIM: 167790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPINK1 PubMed Links

Gene Details

Gene
SPINK1
Gene Name
serine peptidase inhibitor, Kazal type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003122.4 Intron NP_003113.2

View Full Product Details