Product Details

SNP ID
rs141209810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:14706846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCAAAAACATGCTTTCCTACG[A/G]TTTGAGTGCTTTATATGAGAGCCAT
Phenotype
MIM: 605145 MIM: 615712
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKH PubMed Links

Gene Details

Gene
ANKH
Gene Name
ANKH inorganic pyrophosphate transport regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054027.4 6161 UTR 3 NP_473368.1
XM_011514067.1 6161 Intron XP_011512369.1
XM_017009644.1 6161 UTR 3 XP_016865133.1
Gene
LOC100130744
Gene Name
uncharacterized LOC100130744
There are no transcripts associated with this gene.

Gene
OTULIN
Gene Name
OTU deubiquitinase with linear linkage specificity
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138348.4 6161 Intron NP_612357.4
XM_011514151.2 6161 Intron XP_011512453.1
XM_011514152.2 6161 Intron XP_011512454.1
XM_011514154.2 6161 Intron XP_011512456.1
XM_017010015.1 6161 Intron XP_016865504.1

View Full Product Details