Product Details
- SNP ID
-
rs141365967
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:178613482 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCAAGTGTACTTACAATTTATTG[A/G]CTGGCAGATCTGATACGCCATCTGC
- Phenotype
-
MIM: 607969
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLK4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs566021338] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLK4
- Gene Name
- CDC like kinase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020666.2 |
925 |
Missense Mutation |
CCA,TCA |
P273S |
NP_065717.1 |
View Full Product Details