Product Details
- SNP ID
-
rs141463827
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55107850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCAATAACCAGGGAAGAGCCAA[C/T]AAAACAGATAATGCTTGTTAAGAAA
- Phenotype
-
MIM: 140050
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDC20B
PubMed Links
Gene Details
- Gene
- CDC20B
- Gene Name
- cell division cycle 20B
There are no transcripts associated with this gene.
- Gene
- GZMA
- Gene Name
- granzyme A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006144.3 |
309 |
Missense Mutation |
ACA,ATA |
T91I |
NP_006135.1 |
View Full Product Details