Product Details
- SNP ID
-
rs141510669
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:178135256 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCTCCCGGGCTCAAGCAGTCCTC[C/T]GTCCCACCTCAGCCTTCTGAGGAGC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
N4BP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10060446] are located under a probe and SNP(s) [rs72536798] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- N4BP3
- Gene Name
- NEDD4 binding protein 3
There are no transcripts associated with this gene.
- Gene
- RMND5B
- Gene Name
- required for meiotic nuclear division 5 homolog B
View Full Product Details