Product Details

SNP ID
rs141748668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:109691159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTGTTTGTGGCAGACAGGGGG[A/C]ACCGAGTGGGGAAGCGCGTGTTATA
Phenotype
MIM: 154582
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAN2A1 PubMed Links

Gene Details

Gene
MAN2A1
Gene Name
mannosidase alpha class 2A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002372.3 Intron NP_002363.2
XM_011543395.2 Intron XP_011541697.1
XM_017009472.1 Intron XP_016864961.1

View Full Product Details