Product Details

SNP ID
rs141936903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:153491368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGTGTGTGTTTTCACGTGTGT[A/G]ATTATATATTACATATGCACATATA
Phenotype
MIM: 138248
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIA1 PubMed Links

Gene Details

Gene
GRIA1
Gene Name
glutamate ionotropic receptor AMPA type subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000827.3 Intron NP_000818.2
NM_001114183.1 Intron NP_001107655.1
NM_001258019.1 Intron NP_001244948.1
NM_001258020.1 Intron NP_001244949.1
NM_001258021.1 Intron NP_001244950.1
NM_001258022.1 Intron NP_001244951.1
NM_001258023.1 Intron NP_001244952.1
XM_011537635.2 Intron XP_011535937.1
XM_017009392.1 Intron XP_016864881.1
XM_017009393.1 Intron XP_016864882.1

View Full Product Details