Product Details

SNP ID
rs141965109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178946981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAGGACACCCCTGGAGGCTTCT[C/G]TCTTGGTAAGAATCATGTGTGTGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF454 PubMed Links

Gene Details

Gene
ZNF454
Gene Name
zinc finger protein 454
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178089.2 606 Missense Mutation TCT,TGT S82C NP_001171560.1
NM_001178090.2 606 Missense Mutation TCT,TGT S82C NP_001171561.1
NM_001323306.1 606 Missense Mutation TCT,TGT S82C NP_001310235.1
NM_001323307.1 606 Missense Mutation TCT,TGT S82C NP_001310236.1
NM_001323308.1 606 Missense Mutation TCT,TGT S82C NP_001310237.1
NM_001323309.1 606 Missense Mutation TCT,TGT S82C NP_001310238.1
NM_001323310.1 606 Missense Mutation TCT,TGT S82C NP_001310239.1
NM_182594.3 606 Missense Mutation TCT,TGT S82C NP_872400.2

View Full Product Details