Product Details

SNP ID
rs142191083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:74628469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGACTAGCACTAATACCTACGA[C/T]GGCACATGCTTAGTAAACAAGCACT
Phenotype
MIM: 605173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENC1 PubMed Links

Gene Details

Gene
ENC1
Gene Name
ectodermal-neural cortex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256574.1 4665 UTR 3 NP_001243503.1
NM_001256575.1 4665 UTR 3 NP_001243504.1
NM_001256576.1 4665 UTR 3 NP_001243505.1
NM_003633.3 4665 UTR 3 NP_003624.1
XM_011543696.2 4665 UTR 3 XP_011541998.1
XM_011543697.2 4665 UTR 3 XP_011541999.1

View Full Product Details