Product Details

SNP ID
rs142284487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:865502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGCCGCGCTCCGCCTGTGCTTC[A/G]TGCAGGTCCTGCAGGAGCTTCGTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD9 PubMed Links

Gene Details

Gene
BRD9
Gene Name
bromodomain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009877.2 1599 Silent Mutation CAC,CAT H482H NP_001009877.2
NM_001317951.1 1599 Silent Mutation CAC,CAT H439H NP_001304880.1
NM_023924.4 1599 Silent Mutation CAC,CAT H535H NP_076413.3
XM_006714494.3 1599 Intron XP_006714557.1
XM_017009780.1 1599 Intron XP_016865269.1
Gene
ZDHHC11
Gene Name
zinc finger DHHC-type containing 11
There are no transcripts associated with this gene.

View Full Product Details