Product Details

SNP ID
rs142314227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154889321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATGAACTGCTTTAACTCTTAC[A/C]TTAATGCTCTCAGGAAATTTCGCCA
Phenotype
MIM: 607005
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GEMIN5 PubMed Links

Gene Details

Gene
GEMIN5
Gene Name
gem nuclear organelle associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252156.1 4439 Missense Mutation AAG,AAT K1452N NP_001239085.1
NM_015465.4 4439 Missense Mutation AAG,AAT K1453N NP_056280.2

View Full Product Details