Product Details

SNP ID
rs142501162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132674125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCGGCAACTTTGTCCACGGACA[C/T]AAGTGCGATATCACCTTACAGGAGA
Phenotype
MIM: 147780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL4 PubMed Links

Gene Details

Gene
IL4
Gene Name
interleukin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000589.3 140 Silent Mutation CAC,CAT H25H NP_000580.1
NM_172348.2 140 Silent Mutation CAC,CAT H25H NP_758858.1
Gene
LOC105379176
Gene Name
uncharacterized LOC105379176
There are no transcripts associated with this gene.

View Full Product Details