Product Details

SNP ID
rs142724357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55160244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTTCCGGGCCCAGAGCAAAG[C/G]TATTTGCAGTTTTGCTGTCTATAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC20B PubMed Links

Gene Details

Gene
CDC20B
Gene Name
cell division cycle 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145734.2 127 Intron NP_001139206.2
NM_001170402.1 127 Intron NP_001163873.1
NM_152623.2 127 Intron NP_689836.2
XM_011543218.2 127 Intron XP_011541520.1
Gene
GPX8
Gene Name
glutathione peroxidase 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008397.3 127 Missense Mutation CTA,GTA L18V NP_001008398.2
NM_001306197.1 127 Intron NP_001293126.1
NM_001306198.1 127 Missense Mutation CTA,GTA L18V NP_001293127.1
NM_001306201.1 127 Intron NP_001293130.1
XM_006714631.2 127 Missense Mutation CTA,GTA L18V XP_006714694.1

View Full Product Details