Product Details

SNP ID
rs142766405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34935755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTACTTAAAGGTGGGTTTGATGG[C/G]GAATATCAAGATGACAGCTTAGATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC21 PubMed Links

Gene Details

Gene
DNAJC21
Gene Name
DnaJ heat shock protein family (Hsp40) member C21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012339.2 464 Silent Mutation GGC,GGG G79G NP_001012339.2
NM_194283.3 464 Silent Mutation GGC,GGG G79G NP_919259.3
XM_005248249.4 464 Silent Mutation GGC,GGG G166G XP_005248306.2
XM_005248250.3 464 Silent Mutation GGC,GGG G166G XP_005248307.2
XM_011513965.2 464 Silent Mutation GGC,GGG G166G XP_011512267.2
XM_011513966.2 464 Silent Mutation GGC,GGG G166G XP_011512268.2

View Full Product Details