Product Details

SNP ID
rs143209032
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141319221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGAATTCCCATAACCAGCTGA[C/T]TGGTTCCTTCATTTTCCTGGTGCTG
Phenotype
MIM: 600573
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TAF7 PubMed Links

Gene Details

Gene
TAF7
Gene Name
TATA-box binding protein associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005642.2 1564 Missense Mutation AAT,AGT N275S NP_005633.2

View Full Product Details