Product Details

SNP ID
rs143278360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:119452596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCATGGGCTCACCGCTGAGGTT[C/T]GACGGGCGGGTGGTACTGGTCACCG
Phenotype
MIM: 601860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B4 PubMed Links

Gene Details

Gene
HSD17B4
Gene Name
hydroxysteroid 17-beta dehydrogenase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000414.3 154 Silent Mutation TTC,TTT F7F NP_000405.1
NM_001199291.2 154 UTR 5 NP_001186220.1
NM_001199292.1 154 Silent Mutation TTC,TTT F7F NP_001186221.1
NM_001292027.1 154 UTR 5 NP_001278956.1
NM_001292028.1 154 UTR 5 NP_001278957.1

View Full Product Details